Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-2,6-bis(hydroxymethyl)-4-pyrone 98.0+%, TCI America™
SDP

Catalog No. H10191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10191G 1 g
H10195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10191G Supplier TCI America Supplier No. H10191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2029-49-4
Molecular Formula C7H8O5
Molecular Weight (g/mol) 172.14
MDL Number MFCD00110732
InChI Key YZUXYQQFVNGYCA-UHFFFAOYSA-N
PubChem CID 2747691
IUPAC Name 3-hydroxy-2,6-bis(hydroxymethyl)-4H-pyran-4-one
SMILES OCC1=CC(=O)C(O)=C(CO)O1

Specifications

Melting Point 149°C
Quantity 1 g
Formula Weight 172.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2,6-bis(hydroxymethyl)-4-pyrone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.