Learn More
3-Hydroxy-2-butanone, monomer + dimer, 95%
Description
3-Hydroxy-2-butanone is a chemical used in food flavoring and fragrances. It acts as an intermediate of butanediol cycle in microorganisms. It is used as an aroma carrier in the preparation of flavors and essences.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Hydroxy-2-butanone is a chemical used in food flavoring and fragrances. It acts as an intermediate of butanediol cycle in microorganisms. It is used as an aroma carrier in the preparation of flavors and essences.
Solubility
Miscible with water, propylene glycol and alcohol. Slightly miscible with ether, petroleum ether. Immiscible with fatty oils and vegetable oil.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Density | 1.013 |
| Boiling Point | 148°C |
| Flash Point | 41°C (106°F) |
| Odor | Characteristic |
| Quantity | 1000 g |
| UN Number | UN2621 |
| Beilstein | 385636 |
| Merck Index | 14,64 |
| Solubility Information | Miscible with water,propylene glycol and alcohol. Slightly miscible with ether,petroleum ether. Immiscible with fatty oils and vegetable oil. |
| Formula Weight | 88.11 |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319 |
| Hazard Statement | GHS H Statement H226-H315-H319 Flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ACETYL METHYL CARBINOL |
| EINECSNumber | 208-174-1 |
| RTECSNumber | EL8790000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.