Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-3-methyl-2-butanone 95.0+%, TCI America™
SDP

Catalog No. H0471100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0471100ML 100 mL
H047125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0471100ML Supplier TCI America Supplier No. H0471100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 115-22-0
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004460
InChI Key BNDRWEVUODOUDW-UHFFFAOYSA-N
Synonym 3-hydroxy-3-methyl-2-butanone, 3-methylacetoin, methylacetoin, 2-butanone, 3-hydroxy-3-methyl, dimethylacetylcarbinol, 2-hydroxy-2-methyl-3-butanone, 2-methyl-2-hydroxybutan-3-one, 1-hydroxy-1-methylethyl methyl ketone, 3-hydroxy-3-methyl-butan-2-one, 3-hydroxy-3-ethylbutanone
PubChem CID 8261
IUPAC Name 3-hydroxy-3-methylbutan-2-one
SMILES CC(=O)C(C)(C)O

Specifications

Color Yellow
Boiling Point 138°C
Quantity 100 mL
UN Number 1993
Formula Weight 102.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Hydroxy-3-methyl-2-butanone
EINECSNumber (2)-0594
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.