Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4,5-dimethoxybenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. H11791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11791G 1 g
H11795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11791G Supplier TCI America Supplier No. H11791G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29865-90-5
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00010442
InChI Key NVLTWXMZECWWPC-UHFFFAOYSA-N
Synonym 5-Hydroxyveratraldehyde, 5-Hydroxy-3,4-dimethoxybenzaldehyde
PubChem CID 141513
IUPAC Name 3-hydroxy-4,5-dimethoxybenzaldehyde
SMILES COC1=CC(C=O)=CC(O)=C1OC

Specifications

Melting Point 66°C
Boiling Point 180°C
Quantity 1 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-4,5-dimethoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.