Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4-methoxyaniline 98.0+%, TCI America™
SDP

Catalog No. H063425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H063425G 25 g
H06345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H063425G Supplier TCI America Supplier No. H063425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1687-53-2
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00010222
InChI Key BLQFHJKRTDIZLX-UHFFFAOYSA-N
Synonym 3-hydroxy-4-methoxyaniline, 5-aminoguaiacol, 2-methoxy-5-aminophenol, phenol, 5-amino-2-methoxy, 5-amino-2-methoxy-phenol, pubchem3112, acmc-209dyt, 3-hydroxy4-methoxyaniline, 5-amino-2-methoxy phenol, 3-hydroxy-4-methoxylaniline
PubChem CID 74314
IUPAC Name 5-amino-2-methoxyphenol
SMILES COC1=C(C=C(C=C1)N)O

Specifications

Melting Point 130°C
Quantity 25 g
Formula Weight 139.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-4-methoxyaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.