Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-5-methylisoxazole 98.0+%, TCI America™
SDP

Catalog No. H134825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H134825G 25 g
H13485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H134825G Supplier TCI America Supplier No. H134825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10004-44-1
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.089
MDL Number MFCD00144468
InChI Key KGVPNLBXJKTABS-UHFFFAOYSA-N
Synonym hymexazol, 3-hydroxy-5-methylisoxazole, hymexazole, 5-methylisoxazol-3-ol, 5-methylisoxazol-3 2h-one, hydroxyisoxazole, tachigaren, itachigarden, 5-methyl-3-hydroxyisoxazole, 5-methyl-3 2h-isoxazolone
PubChem CID 24781
ChEBI CHEBI:5827
IUPAC Name 5-methyl-1,2-oxazol-3-one
SMILES CC1=CC(=O)NO1

Specifications

Melting Point 86°C
Quantity 25 g
Formula Weight 99.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-5-methylisoxazole
RTECSNumber NY2932000
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.