Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-5-methylpyridine, 97%, Thermo Scientific™

Catalog No. AC448740050
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC448740050 5 g
AC448742500 250 mg
AC448740010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC448740050 Supplier Thermo Scientific Chemicals Supplier No. 448740050
Add to Cart

Chemical Identifiers

CAS 42732-49-0
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
InChI Key RYJNCIGFPWGVPA-UHFFFAOYSA-N
Synonym 3-hydroxy-5-picoline, 3-hydroxy-5-methylpyridine, 5-methyl-3-hydroxypyridine, 5-hydroxy-3-picoline, 5-methyl-3-pyridinol, 5-methyl-3-pyridol, pubchem6629, 5-methyl-3-hydroxy-pyridine, 3-pyridinol, 5-methyl, 3-hydroxy-5-methylpyridine 250mg
PubChem CID 224753
IUPAC Name 5-methylpyridin-3-ol
SMILES CC1=CC(=CN=C1)O

Specifications

Melting Point 136.0°C to 140.0°C
Assay Percent Range 96.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: slightly soluble
Formula Weight 109.13
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-5-methylpyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.