Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-alpha-methylbenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. H14331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14331G 1 g
H14335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14331G Supplier TCI America Supplier No. H14331G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2415-09-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00238621
InChI Key COJRWHSKVYUZHQ-UHFFFAOYSA-N
Synonym 1-(3-Hydroxyphenyl)ethanol, 3-(1-Hydroxyethyl)phenol
PubChem CID 13542886
IUPAC Name 3-(1-hydroxyethyl)phenol
SMILES CC(C1=CC(=CC=C1)O)O

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 1 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-alpha-methylbenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.