Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxybenzylamine, 96%, Thermo Scientific Chemicals

Catalog No. aah3570806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3570806 5 g
AAH3570803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3570806 Supplier Thermo Scientific Chemicals Supplier No. H3570806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 73604-31-6 | Molecular Formula: C7H9NO | Molecular Weight: 123.16 g/mol

3-Hydroxybenzylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybenzylamine is used as pharmaceutical intermediate.

Solubility
Soluble in water and ethanol.

Notes
Air sensitive. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73604-31-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00798977
InChI Key JNZYADHPGVZMQK-UHFFFAOYSA-N
Synonym 3-aminomethyl phenol, 3-hydroxybenzylamine, phenol, 3-aminomethyl, 3-hydroxybenzylamine, 3-hydroxyphenyl methylamine, 3-aminomethyl-phenol, 3-hydroxy-benzylamine, 3-hydroxyphenyl methylamine
PubChem CID 735894
IUPAC Name 3-(aminomethyl)phenol
SMILES NCC1=CC(O)=CC=C1

Specifications

Melting Point 170°C to 173°C
Quantity 5 g
UN Number UN3259
Sensitivity Air sensitive
Solubility Information Soluble in water and ethanol.
Formula Weight 123.15
Percent Purity 97%
Chemical Name or Material 3-Hydroxybenzylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.