Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Hydroxybutyric acid, 98%

Manufacturer:  Alfa Aesar™ H3042614

Quantity 25g

 View more versions of this product

Catalog No. AAH3042614


  Check Availability
Add to cart

Specifications

Description

Applications
3-Hydroxybutyric acid is used in enantioselective synthesis of D-(-)-3-hydroxybutyric acid by escherichia coli. It forms films of random copolymer with (R)-3-hydroxypentanoic acid, (R)-3-hydroxyhexanoic acid, 4-hydroxybutyric acid, 6-hydroxyhexanoic acid and (S,S)-lactide by the melt-crystallized method.

Solubility
Miscible with water, alcohol and ether.

Notes
Hygroscopic. Incompatible with acids and bases.
Description & Specifications

Chemical Identifiers

300-85-6
104.1
14,4816
C4H8O3
MFCD00004546
3-Hydroxybutyric acid

Specifications

Odorless
25g
C4H8O3
MFCD00004546
3-Hydroxybutyric acid
36° to 39°C
300-85-6
104.1
14,4816
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit