Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxydiphenylamine 98.0+%, TCI America™
SDP

Catalog No. A047625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A047625G 25 g
A0476500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A047625G Supplier TCI America Supplier No. A047625G

Chemical Identifiers

CAS 101-18-8
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00002262
InChI Key NDACNGSDAFKTGE-UHFFFAOYSA-N
Synonym 3-hydroxydiphenylamine, 3-phenylamino phenol, m-anilinophenol, phenol, 3-phenylamino, 3-hydroxy-n-phenylaniline, m-hydroxydiphenylamine, 3-phenylaminophenol, phenol, m-anilino, unii-e39q946go3, ccris 4658
PubChem CID 7546
IUPAC Name 3-anilinophenol
SMILES C1=CC=C(C=C1)NC2=CC(=CC=C2)O

Specifications

Melting Point 80°C
Boiling Point 340°C
Quantity 25 g
Formula Weight 185.23
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxydiphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.