Learn More
Eurofins Discoverx 3-Hydroxyoctanoic acid, 5 mg. (92-1189)
Supplier:  Eurofins Discoverx 921189
Encompass_Preferred
3-Hydroxyoctanoic acid is a signaling chemical emitted by orchids that may also be a marker for primary defects of the β-hydroxy fatty acid metabolism. A polymer of 3-hydroxyoctanoic acid produces a carbon and energy reserve form for bacteria. Also used to produce polyhydroxyalkanoates used in medical devices and tissue engineering applications. Provided as 5mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9717
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.