Learn More
Medchemexpress LLC 3-Hydroxystearic aci 10mg | 17773-30-7 | 300.48 g/mol | C18H36O3 | 10 MG

Supplier: Medchemexpress LLC HY11613310MG
3-Hydroxystearic acid is a hydroxylated C18 saturated fatty acid supplied as a white to off-white solid for research and analytical applications. It is commonly used as an analytical standard and biochemical reagent.
- Purity: 98.25% (HPLC)
- CAS number: 17773-30-7
- Molecular formula: C18H36O3; molecular weight: 300.48 g/mol
- Appearance: white to off-white solid
- Storage: powder -20°C (long term) or 4°C (short term); in solvent -80°C recommended
- Typical use: analytical standard, biochemical research, and method development
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.