Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxyxanthen-9-one 98.0+%, TCI America™
SDP

Catalog No. H12691G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12691G 1 g
H12695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12691G Supplier TCI America Supplier No. H12691G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3722-51-8
Molecular Formula C13H8O3
Molecular Weight (g/mol) 212.204
MDL Number MFCD00604748
InChI Key XCJHDJAODLKGLG-UHFFFAOYSA-N
Synonym sieber linker, 3-hydroxy-9h-xanthen-9-one, 3-hydroxy-xanthen-9-one, 3-hydroxyxanthone, 9h-xanthen-9-one, 3-hydroxy, sieber linker;, xanthen-9-one, 3-hydroxy, pubchem12819, acmc-209irq, 3-hydroxy-9-xanthenone
PubChem CID 5376013
IUPAC Name 3-hydroxyxanthen-9-one
SMILES C1=CC=C2C(=C1)C(=O)C3=C(O2)C=C(C=C3)O

Specifications

Melting Point 243°C
Color White-Yellow
Quantity 1 g
Formula Weight 212.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxyxanthen-9-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.