Learn More
Sigma Organic Chemistry 3-HYDy-6H-benzo cchrom | 250mg | 1139-83-9 | MFCD00034338
Supplier: Sigma Organic Chemistry OTV000001250MG

3-HYDy-6H-benzo cchrom , 250mg
About this item:
CAS #: 1139-83-9
MDL #: MFCD00034338
Chemical Formula: C13H8O3
Molecular Weight: 212.2
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.