Learn More
Scientific Industries 3-INCH PLATFORM
Supplier:  Scientific Industries 0K0500901
Encompass_Preferred
Replacement 3-inch (76mm) platform without rubber cover (Part No. 580-2013-00). This may be used on the Vortex-Genie 1, Vortex-Genie Pulse and the Vortex-Genie 2 family of mixers.
Catalog No. 50-728-061
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.