Learn More
Abcam 3-Indole Acetic Acid, Solid, 99 Percent Purity, 25 g
Description
- Molecular Weight: 175.19 g/mol
- Purity: >99%
- Form: Solid
- CAS Number: 87-51-4
- Description: Natural phytohormone (plant auxin) synthesized from tryptophan that induces plant cell elongation and division.
- Applications: Used as a supplement in media such as Murashige and Skoog media and Gamborg's B5 media.
- Storage: Store at -20°C.
- Shipping: Ambient - Can Ship with Ice.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | 3-Indole Acetic Acid |
| Shipping Condition | Ambient |
| Form | Solid |
| Content And Storage | Store at -20°C. |
| Quantity | 25 g |
| For Use With (Application) | Plant cell culture, plant growth regulator |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.