Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Indolemethanol 98.0+%, TCI America™
SDP

Catalog No. I049625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I049625G 25 g
I04965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I049625G Supplier TCI America Supplier No. I049625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 700-06-1
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00005632
InChI Key IVYPNXXAYMYVSP-UHFFFAOYSA-N
Synonym indole-3-carbinol, 3-indolemethanol, 1h-indol-3-yl methanol, indole-3-methanol, 1h-indole-3-methanol, 3-hydroxymethylindole, 3-indolylcarbinol, indinol, 3-indole methanol, indole 3 carbinol
PubChem CID 3712
ChEBI CHEBI:24814
IUPAC Name 1H-indol-3-ylmethanol
SMILES C1=CC=C2C(=C1)C(=CN2)CO

Specifications

Quantity 25 g
Formula Weight 147.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Indolemethanol
RTECSNumber NL9483000
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.