Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodo-1,2,4,5-tetramethylbenzene 98.0+%, TCI America™
SDP

Catalog No. T09921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09921G 1 g
T09925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09921G Supplier TCI America Supplier No. T09921G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2100-25-6
Molecular Formula C10H13I
Molecular Weight (g/mol) 260.12
MDL Number MFCD00013705
InChI Key MCRSYYLHVQGFNR-UHFFFAOYSA-N
Synonym 2,3,5,6-tetramethyliodobenzene, iododurene, 3-iododurene, benzene, 3-iodo-1,2,4,5-tetramethyl, 1-iodo-2,3,5,6-tetramethylbenzene, 3-iodo-1,2,4,5-tetramethyl-benzene, acmc-209fgl, maybridge1_002808, 3-iodo-1,4,5-tetramethylbenzene, iodo-2,3,5,6-tetramethylbenzene
PubChem CID 75011
IUPAC Name 3-iodo-1,2,4,5-tetramethylbenzene
SMILES CC1=CC(C)=C(C)C(I)=C1C

Specifications

Melting Point 80°C
Color White-Yellow
Boiling Point 290°C
Quantity 1 g
Formula Weight 260.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodo-1,2,4,5-tetramethylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.