Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodo-6-nitroindazole 98.0+%, TCI America™
SDP

Catalog No. I10041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10041G 1 g
I10045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10041G Supplier TCI America Supplier No. I10041G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 70315-70-7
Molecular Formula C7H4IN3O2
Molecular Weight (g/mol) 289.03
MDL Number MFCD07781652
InChI Key GZCGNGLOCQEDMT-UHFFFAOYSA-N
PubChem CID 12475306
IUPAC Name 3-iodo-6-nitro-2H-indazole
SMILES [O-][N+](=O)C1=CC2=NNC(I)=C2C=C1

Specifications

Melting Point 265°C
Quantity 1 g
Formula Weight 289.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodo-6-nitroindazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.