Learn More
Sigma Organic Chemistry 3-Iodoaniline | 25G | 626-01-7 | MFCD00007781
Supplier: Sigma Organic Chemistry I720925G

3-Iodoaniline, 25G
About This Item:
Linear Formula: IC6H4NH2
Molecular Weight: 219.02
CAS Number: 626-01-7
MDL Number: MFCD00007781
UNSPSC Code: 12352100
Purity: 0.98
Boiling Point: 145-146 C/15 mmHg (lit.)
Melting Point: 21-24 C (lit.)
Density: 1.821 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.682 (lit.)
Storage: 2-8C
EINECS Number: 210-922-7
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.