Learn More
3-Iodoanisole, 98%
Description
3-Iodoanisole is used to produce 3,3'-Dimethoxy-biphenyl. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Iodoanisole is used to produce 3,3′-Dimethoxy-biphenyl. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in chloroform, alcohol, ether. Insoluble in water.
Notes
Light Sensitive. Incompatible with light and oxidizing agents. Store in the dark.
Specifications
Specifications
| Density | 1.78 |
| Boiling Point | 243°C to 244°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.613 |
| Quantity | 25 g |
| Beilstein | 1858896 |
| Sensitivity | Light sensitive |
| Solubility Information | Soluble in chloroform,alcohol,ether. Insoluble in water. |
| Formula Weight | 234.04 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H315-H320-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 212-173-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.