Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. I005225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I005225G 25 g
I00525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I005225G Supplier TCI America Supplier No. I005225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 618-51-9
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.019
MDL Number MFCD00002496
InChI Key KVBWBCRPWVKFQT-UHFFFAOYSA-N
Synonym m-iodobenzoic acid, benzoic acid, 3-iodo, 3-iodo-benzoic acid, benzoic acid, m-iodo, 3-iodobenzoicacid, 3-carboxyiodobenzene, 3-iodo benzoic acid, pubchem3925, acmc-209mwv, meta-carboxy iodo-benzene
PubChem CID 12060
IUPAC Name 3-iodobenzoic acid
SMILES C1=CC(=CC(=C1)I)C(=O)O

Specifications

Melting Point 187°C
Quantity 25 g
Formula Weight 248.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.