Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodobenzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. I07201G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07201G 1 g
I07205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07201G Supplier TCI America Supplier No. I07201G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1711-10-0
Molecular Formula C7H4ClIO
Molecular Weight (g/mol) 266.462
MDL Number MFCD02093934
InChI Key DTELTOREECFDBC-UHFFFAOYSA-N
Synonym 3-iodobenzoylchloride, m-iodobenzoyl chloride, benzoyl chloride, 3-iodo, 3-iodo benzoyl chloride, 3-iodo-benzoyl chloride, benzoylchloride,3-iodo, benzoyl chloride,3-iodo, acmc-1bql4, 3-iodobenzoic acid chloride, 3-iodobenzoyl chloride, 97+%
PubChem CID 74378
IUPAC Name 3-iodobenzoyl chloride
SMILES C1=CC(=CC(=C1)I)C(=O)Cl

Specifications

Melting Point 25°C
Boiling Point 146°C
Quantity 1 g
UN Number 3261
Formula Weight 266.46
Percent Purity ≥98.0% (GC,T)
Physical Form Crystals
Chemical Name or Material 3-Iodobenzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.