Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodobenzyl Bromide 97.0+%, TCI America™
SDP

Catalog No. I060325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I060325G 25 g
I06035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I060325G Supplier TCI America Supplier No. I060325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49617-83-6
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.933
InChI Key BACZSVQZBSCWIG-UHFFFAOYSA-N
Synonym 3-iodobenzyl bromide, 1-bromomethyl-3-iodobenzene, alpha-bromo-3-iodotoluene, 3-iodobenzylbromide, m-iodobenzyl bromide, benzene, 1-bromomethyl-3-iodo, m-iodobenzylbromide, 3-iodo-benzylbromide, 3-iodo-benzyl bromide, meta-iodobenzyl bromide
PubChem CID 2759361
IUPAC Name 1-(bromomethyl)-3-iodobenzene
SMILES C1=CC(=CC(=C1)I)CBr

Specifications

Melting Point 52°C
Quantity 25 g
UN Number 3261
Formula Weight 296.93
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodobenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.