Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodobenzylamine hydrochloride, 98+%, Thermo Scientific™

Catalog No. AAL0562203
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0562203 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0562203 Supplier Thermo Scientific Chemicals Supplier No. L0562203

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3718-88-5
Molecular Formula C7H9ClIN
Quantity 1g
MDL Number MFCD00012857
Beilstein 5352277
Synonym 3-iodobenzylamine hydrochloride, 3-iodophenyl methanamine hydrochloride, 3-iodobenzylamine hcl, 3-iodobenzylaminehydrochloride, benzenemethanamine, 3-iodo-, hydrochloride, 1-3-iodophenyl methanamine hydrochloride, pubchem3302, acmc-1cmc7, 3-iodobenzylamine hcl salt, ksc491e3h
InChI Key PYFDZOCGFHIRST-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)I)CN.Cl
IUPAC Name (3-iodophenyl)methanamine;hydrochloride
Molecular Weight (g/mol) 269.51
PubChem CID 2723861
Formula Weight 269.51
Percent Purity ≥98%
Chemical Name or Material 3-Iodobenzylamine hydrochloride
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.