Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Isopropylbenzeneboronic acid, 99%

Catalog No. AAL1553003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1553003 1 g
AAL15530MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1553003 Supplier Thermo Scientific Chemicals Supplier No. L1553003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 216019-28-2 | C9H13BO2 | 164.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216019-28-2
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.01
MDL Number MFCD01074634
InChI Key QSWLFBMVIGQONC-UHFFFAOYSA-N
Synonym 3-isopropylphenylboronic acid, 3-isopropylbenzeneboronic acid, 3-cumylboronic acid, 3-isopropylphenyl boronic acid, 3-isopropylphebylboronic acid, 3-propan-2-yl phenyl boranediol, boronic acid, 3-1-methylethyl phenyl, boronic acid, b-3-propylphenyl, 3-propan-2-yl phenyl boronic acid, 3-propan-2-ylphenyl boronic acid
PubChem CID 2773478
IUPAC Name (3-propan-2-ylphenyl)boronic acid
SMILES CC(C)C1=CC=CC(=C1)B(O)O

Specifications

Melting Point ∼75°C to 80°C
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 3090001
Formula Weight 164.01
Percent Purity 99%
Chemical Name or Material 3-Isopropylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.