Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Mercapto-1-propanol 97.0+%, TCI America™
SDP

Catalog No. M120625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M120625G 25 g
M12065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. M120625G Supplier TCI America Supplier No. M120625G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 19721-22-3
Molecular Formula C3H8OS
Molecular Weight (g/mol) 92.156
MDL Number MFCD00192262
InChI Key SHLSSLVZXJBVHE-UHFFFAOYSA-N
Synonym 3-mercapto-1-propanol, 1-propanol, 3-mercapto, 3-mercaptopropanol, 3-hydroxypropanethiol, 3-hydroxypropyl mercaptan, 3-mecapto-1-propanol, 3-mecaptopropan-1-ol, 1-mercapto-3-propanol, 3-hydroxypropylmercaptan, 3-mercaptopropan-1-ol
PubChem CID 88211
IUPAC Name 3-sulfanylpropan-1-ol
SMILES C(CO)CS

Specifications

Color Yellow
Boiling Point 90°C
Quantity 25 g
UN Number 2810
Formula Weight 92.16
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Mercapto-1-propanol
RTECSNumber UB7630000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.