Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Methanesulfonyl)phenylboronic acid, 98%, Thermo Scientific™

Catalog No. AC454240050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454240050 5 g
AC454240010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454240050 Supplier Thermo Scientific Chemicals Supplier No. 454240050

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 373384-18-0
Molecular Formula C7H9BO4S
Molecular Weight (g/mol) 200.02
InChI Key HZFFUMBZBGETES-UHFFFAOYSA-N
Synonym 3-methylsulfonyl phenylboronic acid, 3-methylsulfonyl phenyl boronic acid, 3-methylsulfonylphenyl boronic acid, 3-methanesulfonyl phenylboronic acid, 3-methanesulfonyl phenyl boronic acid, 3-methanesulfonylphenylboronic acid, boronic acid, 3-methylsulfonyl phenyl, 3-methanesulfonylphenyl boronic acid, 3-methanesulphonyl phenylboronic acid
PubChem CID 2773542
IUPAC Name (3-methylsulfonylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)S(=O)(=O)C)(O)O

Specifications

Melting Point 102.0°C to 110.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 200.02
Percent Purity 98%
Chemical Name or Material 3-(Methanesulfonyl)phenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.