Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-4-methylbenzonitrile 98.0+%, TCI America™
SDP

Catalog No. M19771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19771G 1 g
M19775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19771G Supplier TCI America Supplier No. M19771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3556-60-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD03428590
InChI Key QLJZMAGXHHXXMS-UHFFFAOYSA-N
Synonym 4-Cyano-2-methoxytoluene, 5-Cyano-2-methylanisole
PubChem CID 14752994
IUPAC Name 3-methoxy-4-methylbenzonitrile
SMILES CC1=C(C=C(C=C1)C#N)OC

Specifications

Melting Point 54°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxy-4-methylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.