Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-4-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. M208325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M208325G 25 g
M20835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M208325G Supplier TCI America Supplier No. M208325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5081-36-7
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00007353
InChI Key PWURRRRGLCVBMX-UHFFFAOYSA-N
Synonym 3-methoxy-4-nitrobenzoic acid, 4-nitro-3-methoxybenzoic acid, 4-nitro-m-anisic acid, benzoic acid, 3-methoxy-4-nitro, 3-methoxy-4-nitrobenzoicacid, pubchem3918, acmc-1bn9s, ksc270m6t, 3-methoxy4-nitro-benzoic acid, 3-methoxy-4-nitro-benzoic acid
PubChem CID 78764
SMILES COC1=C(C=CC(=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 235°C
Quantity 25 g
Formula Weight 197.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxy-4-nitrobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.