Learn More
Sigma Organic Chemistry 3-methoxy-isoxazole-5 | 100mg | 13626-59-0 | MFCD07368245
Supplier: Sigma Organic Chemistry CDS005261100MG

3-methoxy-isoxazole-5 , 100mg
About this Item:
CAS #: 13626-59-0
MDL #: MFCD07368245
Molecular Weight: 143.1
UNSPSC Code: 12352200
Chemical Formula: C5H5NO4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.