Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxybenzotrifluoride 97.0+%, TCI America™
SDP

Catalog No. M224325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M224325G 25 g
M22435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M224325G Supplier TCI America Supplier No. M224325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 454-90-0
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.14
MDL Number MFCD00000385
InChI Key XHONYVFDZSPELQ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl anisole, 1-methoxy-3-trifluoromethyl benzene, 3-methoxybenzotrifluoride, 3-trifluoromethylanisole, benzene, 1-methoxy-3-trifluoromethyl, 3-methoxy-1-trifluoromethyl benzene, pubchem2712, 3-trifluoromethylanisol, acmc-1cuj0, m-trifluoromethyl anisole
PubChem CID 96691
IUPAC Name 1-methoxy-3-(trifluoromethyl)benzene
SMILES COC1=CC=CC(=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 g
UN Number 1993
Formula Weight 176.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxybenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.