Learn More
Sigma Organic Chemistry 3-Methoxybutyl acetate | 1G | 4435-53-4 | MFCD00043928
Supplier: Sigma Organic Chemistry CDS0005091G

3-Methoxybutyl acetate , 1G
About this Item:
CAS #: 4435-53-4
MDL #: MFCD00043928
Molecular Weight: 146.18
UNSPSC Code: 12352200
Chemical Formula: C7H14O3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.