Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxyflavone 98.0+%, TCI America™
SDP

Catalog No. M23641G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23641G 1 g
M23645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23641G Supplier TCI America Supplier No. M23641G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7245-02-5
Molecular Formula C16H12O3
Molecular Weight (g/mol) 252.27
MDL Number MFCD00017612
InChI Key ZAIANDVQAMEDFL-UHFFFAOYSA-N
PubChem CID 146013
IUPAC Name 3-methoxy-2-phenylchromen-4-one
SMILES COC1=C(OC2=CC=CC=C2C1=O)C3=CC=CC=C3

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 1 g
Formula Weight 252.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxyflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.