Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-3-Methoxymorphinan hydrochloride, For drug analysis, MilliporeSigma™ Supelco™

Catalog No. 111012434 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2434 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2434 Supplier MilliporeSigma Supelco Supplier No. M18725MG
Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 1087-69-0
Physical Form Powder
Linear Formula C17H23 NO · HCl
MDL Number MFCD02173900
Synonym (+)-cis-1,3,4,9,10,10 A-Hexahydro-6-methoxy-2 H-10,4 A-iminoethanophenanthrene
Solubility Soluble in water
Type Analytical Standard
Industry Type Drug Analysis
Quantity 25 mg
Formula Weight 293.83
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.