Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxyphenyl Isocyanate 97.0+%, TCI America™
SDP

Catalog No. I055125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I055125G 25 g
I05515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I055125G Supplier TCI America Supplier No. I055125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18908-07-1
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00002019
InChI Key NPOVTGVGOBJZPY-UHFFFAOYSA-N
Synonym 3-methoxyphenyl isocyanate, benzene, 1-isocyanato-3-methoxy, isocyanic acid 3-methoxyphenyl ester, 3-isocyanatoanisole, 3-methoxyphenylisocyanate, m-methoxyphenyl isocyanate, 1-isocyanato-3-methoxy-benzene, 3-methoxybenzenisocyanate, m-anisyl isocyanate, m-methoxyphenylisocyanate
PubChem CID 87843
IUPAC Name 1-isocyanato-3-methoxybenzene
SMILES COC1=CC=CC(=C1)N=C=O

Specifications

Color Colorless
Boiling Point 95°C
Quantity 25 g
UN Number 2206
Formula Weight 149.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxyphenyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.