Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M132225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1322-25G 25 g
M1322-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1322-25G Supplier TCI America Supplier No. M132225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10365-98-7
Molecular Formula C7H9BO3
Molecular Weight (g/mol) 151.96
MDL Number MFCD00161359
InChI Key NLLGFYPSWCMUIV-UHFFFAOYSA-N
Synonym 3-methoxyphenyl boronic acid, 3-methoxybenzeneboronic acid, 3-methoxyphenyl boranediol, 3-methoxypenylboronic acid, 3-boronanisole
PubChem CID 2734370
IUPAC Name (3-methoxyphenyl)boronic acid
SMILES COC1=CC=CC(=C1)B(O)O

Specifications

Melting Point 160°C
Quantity 25 g
Formula Weight 151.96
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.