Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxypyridazine 97.0+%, TCI America™
SDP

Catalog No. M20785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20785G 5 g
M20781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20785G Supplier TCI America Supplier No. M20785G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19064-65-4
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD00234098
InChI Key ASFHDLDAWYTMJS-UHFFFAOYSA-N
PubChem CID 292493
IUPAC Name 3-methoxypyridazine
SMILES COC1=NN=CC=C1

Specifications

Boiling Point 87°C
Quantity 5 g
Formula Weight 110.12
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Methoxypyridazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.