Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxythioanisole 98.0+%, TCI America™
SDP

Catalog No. M188025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M188025G 25 g
M18805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M188025G Supplier TCI America Supplier No. M188025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2388-74-1
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.23
MDL Number MFCD03789162
InChI Key IWVQVOXDIOKVBE-UHFFFAOYSA-N
Synonym 3-(Methylthio)anisole
PubChem CID 281182
IUPAC Name 1-methoxy-3-(methylsulfanyl)benzene
SMILES COC1=CC=CC(SC)=C1

Specifications

Color Yellow
Boiling Point 127°C
Quantity 25 g
Formula Weight 154.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxythioanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.