Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-(4-sulfophenyl)-2-pyrazolin-5-one 97.0+%, TCI America™
SDP

Catalog No. M009025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M009025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M009025G Supplier TCI America Supplier No. M009025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89-36-1
Molecular Formula C10H10N2O4S
Molecular Weight (g/mol) 254.26
MDL Number MFCD00020756
InChI Key CWJQQASJVVAXKL-UHFFFAOYSA-N
Synonym 1-(4′C-Sulfophenyl)-3-methyl-5-pyrazolone
PubChem CID 66638
IUPAC Name 4-(3-methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzene-1-sulfonic acid
SMILES CC1=NN(C(=O)C1)C1=CC=C(C=C1)S(O)(=O)=O

Specifications

Color Yellow
Quantity 25 g
UN Number 1759
Formula Weight 254.26
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-1-(4-sulfophenyl)-2-pyrazolin-5-one
EINECSNumber (5)-0349
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.