Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-butanol, 99%, for biochemistry, AcroSeal™

Catalog No. AC412720025
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412720025 2.5 L AcroSeal™ Glass bottle
AC412721000 100 mL AcroSeal™ Glass bottle
AC412720010 1 L AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC412720025 Supplier Thermo Scientific Chemicals Supplier No. 412720025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 123-51-3 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Melting Point -70.0°C
Density 0.8000g/mL
Boiling Point 130.0°C
Flash Point 45°C
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass bottle
Refractive Index 1.4050 to 1.4070
Linear Formula (CH3)2CHCH2CH2OH
Quantity 2.5 L
Beilstein 01,392
Merck Index 15,5241
Specific Gravity 0.8
Solubility Information Solubility in water: 25g/L (20°C). Other solubilities: miscible with alcohol, ether, benzene, oils,, chloroform
Viscosity 4.3 mPa.s (20°C)
Formula Weight 88.15
Percent Purity 99%
Grade Biochemistry
Chemical Name or Material 3-Methyl-1-butanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin
EINECSNumber 204-633-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.