accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3HO

CAS RN 123-51-3

IUPAC Name: 3-methylbutan-1-ol
Synonyms: isoamyl alcohol alcool isoamylique potassium salt isopentyl alcohol isopentyl alcohol (substance) fema number 2057 barium salt isopentyl alcohol magnesium salt isopentyl alcohol lead (2+) salt isopentyl alcohol isopentan-1-ol amylowy alkohol
Molecular Weight (g/mol): 88.15
Molecular Formula: C5H12O
InChi Key: PHTQWCKDNZKARW-UHFFFAOYSA-N
SMILES: CC(C)CCO
Quantity 
  • (8)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (12)
Percent Purity 
  • (11)
  • (1)
  • (4)
  • (2)
  • (1)
  • (4)
  • (6)
  • (1)
Boiling Point 
  • (3)
  • (6)
  • (10)
  • (3)
  • (2)
  • (2)
  • (2)
Grade 
  • (8)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (16)
  • (3)
  • (1)
  • (8)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (12)
  • (11)
  • (1)
  • (4)
  • (2)
  • (1)
  • (4)
  • (6)
  • (1)
  • (3)
  • (6)
  • (10)
  • (3)
  • (2)
  • (2)
  • (2)
  • (8)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
  • (3)
  • (25)
  • (1)