Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-butanol, ACS Reagent, ≥98.5%, Solstice

Catalog No. 60046033 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-033 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-046-033 Supplier Solstice Supplier No. 3200211L

missing translation for 'validMainframeMsg'

ACS Reagent,≥98.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002934
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Melting Point -117°C
Density 0.809g/mL at 25°C
Boiling Point 130°C
Explosion Limit 1.2 to 9% (100°F)
Flash Point 43°C (109.4°F)
Ignition Point 644°F
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2CH2OH
Quantity 1 L
UN Number UN1105
Beilstein 1718835
Vapor Density 3 (vs air)
Vapor Pressure 2mmHg (20°C)
Formula Weight 88.15g/mol
Percent Purity ≥98.5%
Grade ACS Reagent
Chemical Name or Material 3-Methyl-1-Butanol
Show More Show Less
Hazard Statement P210-P261-P280-P304 + P340 + P312-P337 + P313-P403 + P235
Precautionary Statement H226-H315-H319-H332-H335
DOTInformation Class 3 - PG 3 - Pentanols
EINECSNumber 204-633-5
RTECSNumber EL5425000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.