Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2(5H)-furanone 97.0+%, TCI America™
SDP

Catalog No. M10785ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10785ML 5 mL
M10781ML 1 mL
M107825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M10785ML Supplier TCI America Supplier No. M10785ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22122-36-7
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00191545
InChI Key VGHBEMPMIVEGJP-UHFFFAOYSA-N
Synonym 4-Hydroxy-2-methyl-2-butenoic Acid gamma-Lactone
PubChem CID 30945
IUPAC Name 4-methyl-2H-furan-5-one
SMILES CC1=CCOC1=O

Specifications

Color Yellow
Boiling Point 98°C
Quantity 5 mL
Formula Weight 98.10
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2(5H)-furanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.