Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%

Catalog No. p-7023178
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1631706 5 g
AAA1631714 25 g
AAA1631722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1631706 Supplier Thermo Scientific Chemicals Supplier No. A1631706
Only null left
Add to Cart

CAS: 149022-15-1 | C8H10ClN3S | 215.70 g/mol

3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.

Solubility
Soluble in water, dimethyl sulfoxide and methanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149022-15-1
Molecular Formula C8H10ClN3S
Molecular Weight (g/mol) 215.70
MDL Number MFCD00149370
InChI Key OEZPVSPULCMUQB-UHFFFAOYSA-N
Synonym unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride
PubChem CID 9575839
IUPAC Name (Z)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrochloride
SMILES Cl.CN1C(SC2=CC=CC=C12)=NN

Specifications

Melting Point ∼275°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 3571041
Solubility Information Soluble in water,dimethyl sulfoxide and methanol.
Formula Weight 215.71 (Anhydrous)
Percent Purity 97%
Chemical Name or Material 3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 238-428-7
RTECSNumber DL7160200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.