Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-cyclohexen-1-one, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
Packaging:
Glass bottle
1193-18-6
C7H10O
110.16
MFCD00001581
IITQJMYAYSNIMI-UHFFFAOYSA-N
3-methyl-2-cyclohexen-1-one,seudenone,3-methyl-2-cyclohexenone,2-cyclohexen-1-one, 3-methyl,methylcyclohexenone,caswell no. 561ab,fema no. 3360,3-methyl-2-cyclohexene-1-one,epa pesticide chemical code 219700,ghl.pd_mitscher_leg0.699
14511
3-methylcyclohex-2-en-1-one
CC1=CC(=O)CCC1
1193-18-6
C7H10O
110.16
MFCD00001581
IITQJMYAYSNIMI-UHFFFAOYSA-N
3-methyl-2-cyclohexen-1-one,seudenone,3-methyl-2-cyclohexenone,2-cyclohexen-1-one, 3-methyl,methylcyclohexenone,caswell no. 561ab,fema no. 3360,3-methyl-2-cyclohexene-1-one,epa pesticide chemical code 219700,ghl.pd_mitscher_leg0.699
14511
3-methylcyclohex-2-en-1-one
CC1=CC(=O)CCC1
Melting Point -21°C
Flash Point 74°C
Color Yellow to Brown
Boiling Point 199.0°C to 200.0°C
Refractive Index 1.4935 to 1.4955
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 50g
Specific Gravity 0.97
Density 0.9700g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H7(=O)
Beilstein 07, 54
Solubility Information Solubility in water: insoluble
Formula Weight 110.16
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Methyl-2-cyclohexen-1-one
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.