Learn More
Sigma Organic Chemistry 3-Methyl-2-oxo-2 3-dihy | 1G | 62522-63-8 | MFCD03033702
Supplier: Sigma Organic Chemistry CBR014051G

3-Methyl-2-oxo-2 3-dihy , 1G
About this Item:
CAS #: 62522-63-8
MDL #: MFCD03033702
Molecular Weight: 247.66
UNSPSC Code: 12352200
Chemical Formula: C8H6ClNO4S
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.