Learn More
3-Methyl-2-pentanone, 98+%
Description
3-Methyl-2-pentanone is used as a solvent and as an intermediate for syntheses of pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Methyl-2-pentanone is used as a solvent and as an intermediate for syntheses of pharmaceuticals.
Solubility
Soluble in alcohol.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
Specifications
| Density | 0.815 |
| Boiling Point | 118°C to 119°C |
| Flash Point | 12°C (53°F) |
| Refractive Index | 1.4 00 |
| Quantity | 25 g |
| UN Number | UN1224 |
| Beilstein | 635791 |
| Solubility Information | Soluble in alcohol. |
| Formula Weight | 100.16 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H225 |
| Hazard Statement | GHS H Statement H225 Highly flammable liquid and vapour. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S. |
| EINECSNumber | 209-282-1 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.