Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-methyl-2-thienyl)methylamine

Catalog No. CDS015230 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS015230 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS015230 Supplier Sigma Aldrich Supplier No. CDS015230100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-methoxythiophene is a thiophene
  • The intramolecular and intermolecular geometries of crystals of 3-methoxythiophene were studied
  • Spectroscopic studies of bipolarons derived from oligomerized 3-methoxythiophene in solution has been reported
  • The electropolymerization of 3-methoxythiophene on pt and fe electrodes in an aqueous micellar medium containing sodium dodecyl sulfate and 10-3m bithiophene has been reported
  • Thin polymer films of 3-methoxythiophene at the cathode in a direct current discharge have been prepared

Specifications

Molecular Formula C6H9NS
Linear Formula C6H9NS
MDL Number MFCD02093988
Quantity 5 g
Molecular Weight (g/mol) 127.21
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.